tert-Butylphosphonic acid 98%

Danger

Synonyms: Phosphonic acid, (1,1-dimethylethyl)-2-methylprop-2-ylphosphonic acid

AC32152-0010 AC32152-0050
200014-674EA 45 USD
200014-674 103527-614
tert-Butylphosphonic acid 98%
tert-Butylphosphonic acid
Formula: C₄H₁₁O₃P
MW: 138.103 g/mol
Melting Pt: 188…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00012296
CAS Number: 4923-84-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR