Decahydronaphthalene-[D₁₈] (mixture of cis and trans isomers) (98% D) for NMR spectroscopy

Warning

Synonyms: Decahydronaphthalene-[²H₁₈] (mixture of cis and trans isomers)

Packaged in ampoules.

AC32135-0010
200048-936EA 93.5 USD
200048-936
Decahydronaphthalene-[D₁₈] (mixture of cis and trans isomers) (98% D) for NMR spectroscopy
Decahydronaphthalene-[D18] (mixture of cis and trans isomers)
Formula: C₁₀D₁₈
MW: 156.11 g/mol
Density: 1.01 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00044371
CAS Number: 28788-42-3
UN: 1147
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR