Diethyl (hydroxymethyl)phosphonate 97%

Warning

AC32095-0050 AC32095-0250
200006-286EA 52.2 USD
200006-286 103512-354
Diethyl (hydroxymethyl)phosphonate 97%
Diethyl (hydroxymethyl)phosphonate
Formula: C₅H₁₃O₄P
MW: 168.13 g/mol
Boiling Pt: 124…126 °C (3 mmHg)
Density: 1.14 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014418
EINECS: 221-391-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR