Chloro(di-tert-butyl)phosphine 96%

Danger

Synonyms: Di-t-butylchlorophosphineDi-tert-butylchlorophosphine

AC32090-0250 AC32090-0050 AC32090-1000
103527-600EA 254 USD
103527-600 103527-602 200012-172
Chloro(di-tert-butyl)phosphine 96%
Chloro(di-tert-butyl)phosphine
Formula: C₈H₁₈ClP
MW: 180.66 g/mol
Boiling Pt: 69…70 °C (10 mmHg)
Melting Pt: 2…3 °C
Density: 0.95 g/cm³
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00008815
CAS Number: 13716-10-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR