2,4-Dimethylpyrrole 97%

Warning

Synonyms: 2,4-Dimethyl-1H-pyrrole

AC31965-0250 AC31965-0050
103512-344EA 528 USD
103512-344 200004-890
2,4-Dimethylpyrrole 97%
2,4-Dimethylpyrrole
Formula: C₆H₉N
MW: 95.1424 g/mol
Boiling Pt: 165…167 °C
Density: 0.924 g/cm³
Flash Pt: >109 °C
Storage Temperature: Ambient
EINECS: 210-912-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR