Cacodylic acid (dimethylarsinic acid) 98%

Danger

Synonyms: Dimethylarsinic acid (Cacodylic acid)

AC31815-0500 AC31815-2500 AC31815-0050
103527-572EA 47.6 USD
103527-572 103527-574 103527-576
Cacodylic acid (dimethylarsinic acid) 98%
Cacodylic acid (Dimethylarsinic acid)
Formula: (CH₃)₂As(O)OH
MW: 138 g/mol
Melting Pt: 195…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00002095
CAS Number: 75-60-5
EINECS: 200-883-4
UN: 1572
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR