Ethoxycarbonyl isocyanate 90% tech.

Warning

AC31431-0250 AC31431-0050 AC31431-0010
200037-320EA 110 USD
200037-320 103527-514 103527-516
Ethoxycarbonyl isocyanate 90% tech.
Ethoxycarbonyl isocyanate
Formula: C₄H₅NO₃
Boiling Pt: 25 °C (10 mmHg)
Density: 1.11 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00011719
CAS Number: 19617-43-7
UN: 3080
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR