Chloroacetyl isocyanate 90% tech.

Danger

AC31430-0010
103532-360EA 32.2 USD
103532-360
Chloroacetyl isocyanate 90% tech.
Chloroacetyl isocyanate
Formula: C₃H₂ClNO₂
Boiling Pt: 50…55 °C (20 mmHg)
Density: 1.403 g/cm³
Flash Pt: 61 °C
Storage Temperature: Freezer
MDL Number: MFCD00002034
EINECS: 224-715-4
UN: 2922
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR