3'-Isocyanatoacetophenone 97%

Warning

Synonyms: 3-Acetylphenyl isocyanate

AC31410-0010 AC31410-0100
103532-348EA 18.5 USD
103532-348 200035-214
3'-Isocyanatoacetophenone 97%
3'-Isocyanatoacetophenone
Formula: C₉H₇NO₂
Boiling Pt: 155 °C (4 mmHg)
Melting Pt: 33…34 °C
Density: 1.17 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013866
CAS Number: 23138-64-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR