2-(Trifluoromethyl)acrylic acid 98%

Warning

Synonyms: 2-(Trifluoromethyl)propenoic acid

AC31310-0050 AC31310-0010
103512-300EA 159 USD
103512-300 200041-240
2-(Trifluoromethyl)acrylic acid 98%
2-(Trifluoromethyl)acrylic acid
Formula: C₄H₃F₃O₂
MW: 140.06 g/mol
Boiling Pt: 86 °C (760 mmHg)
Melting Pt: 48…52 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00042424
CAS Number: 381-98-6
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR