Perfluorobutane-1-sulfonyl fluoride 92%

Danger

Synonyms: Perfluoro-1-butanesulfonyl fluorideNonafluorobutanesulphonyl fluoridePerfluoro-1-butanesulphonyl fluoride

AC31234-1000 AC31234-0050 AC31234-0250
200013-930EA 68.6 USD
200013-930 103532-326 103532-324
Perfluorobutane-1-sulfonyl fluoride 92%
Perfluorobutane-1-sulphonyl fluoride
Formula: C₄F₁₀O₂S
MW: 302.09
Boiling Pt: 64 °C
Density: 1.68 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007422
CAS Number: 375-72-4
EINECS: 206-792-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR