(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%

Warning

Synonyms: (2R)-(+)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

AC31222-2500 AC31222-0010
200008-216EA 151 USD
200008-216 200021-332
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Boiling Pt: 116…118 °C (1.5 mmHg)
Melting Pt: 46…49 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004184
CAS Number: 20445-31-2
EINECS: 243-829-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR