Poly(methyl acrylate), M.W. ≈ M.W. 40000 (GPC) in toluene

Danger

Synonyms: Acrylic Acid Methyl Ester

AC31069-5000
200058-318EA 66 USD
200058-318
Poly(methyl acrylate), M.W. ≈ M.W. 40000 (GPC) in toluene
Poly(methyl acrylate)

Appearance: Clear liquid

Formula: (C₄H₆O₂)n
Boiling Pt: 110 °C
Density: 1.22 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00084441
CAS Number: 9003-21-8
UN: 1294
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR