Tetraethyl methylenebisphosphonate 97%
|
Warning
|
Synonyms:
Tetraethyl methylenediphosphonate
, Tetraethyl methylenebis(phosphonate)
AC30882-0050
200020-842EA
70.5
USD
200020-842
Tetraethyl methylenebisphosphonate 97%
Tetraethyl methylenebisphosphonate
Formula:
C₉H₂₂O₆P₂ MW: 288.22 g/mol Boiling Pt: 171…174 °C (11 mmHg) Density: 1.16 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039887 EINECS: 216-764-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...