4-Fluorophenyl chloroformate 98%

Danger

Synonyms: 4-Fluorophenyl carbonochloridate

AC30832-0050
200023-802EA 78 USD
200023-802
4-Fluorophenyl chloroformate 98%
4-Fluorophenyl chloroformate
Formula: C₇H₄ClFO₂
MW: 174.5569 g/mol
Flash Pt: 76 °C
Storage Temperature: Refrigerator
UN: 3277
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR