2-Methylene-1,3-propanediol 97%

Warning

Synonyms: 2-Methylenepropane-1,3-diol

AC30859-0010 AC30859-0050
103464-828EA 103 USD
103464-828 200016-694
2-Methylene-1,3-propanediol 97%
2-Methylene-1,3-propanediol
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 93…95 °C (2 mmHg)
Density: 1.08 g/cm³
Flash Pt: <110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075162
CAS Number: 3513-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR