5-Bromo-1,3-benzodioxole 97%

Warning

Synonyms: 4-Bromo-1,2-(methylenedioxy)benzene5-Bromobenzo[d][1,3]dioxole

AC30768-0250
200010-232EA 99.3 USD
200010-232
5-Bromo-1,3-benzodioxole 97%
5-Bromo-1,3-benzodioxole
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 85…86 °C (1 mmHg)
Density: 1.66 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
EINECS: 220-123-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR