Phloretin 98%

Synonyms: 3-(4-Hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)propan-1-one

AC30765-1000 AC30765-5000
200020-384EA 79.1 USD
200020-384 103512-260
Phloretin 98%
Phloretin
Formula: C₁₅H₁₄O₅
MW: 274.28
Melting Pt: 260…262 °C
Storage Temperature: Freezer
EINECS: 200-488-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR