Diethyl fluoromalonate 97%

Danger

Synonyms: 1,3-diethyl 2-fluoropropanedioate

AC30764-0010
200012-040EA 69.5 USD
200012-040
Diethyl fluoromalonate 97%
Diethyl fluoromalonate
Formula: C₇H₁₁FO₄
MW: 178.16 g/mol
Boiling Pt: 121…122 °C (30 mmHg)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00009139
CAS Number: 685-88-1
EINECS: 211-684-7
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR