(Triphenylphosphoranylidene)acetaldehyde 97%

Warning

Synonyms: (Formylmethylene)triphenylphosphorane2-(Triphenylphosphoranylidene)acetaldehyde

AC30749-0050
200027-106EA 44.7 USD
200027-106
(Triphenylphosphoranylidene)acetaldehyde 97%
(Triphenylphosphoranylidene)acetaldehyde
Formula: C₂₀H₁₇OP
MW: 304.33 g/mol
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006994
CAS Number: 2136-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR