4-Isopropylphenylhydrazine hydrochloride 98%

Warning

AC30615-0050
200010-506EA 101 USD
200010-506
4-Isopropylphenylhydrazine hydrochloride 98%
4-Isopropylphenylhydrazine hydrochloride
Formula: C₉H₁₅ClN₂
MW: 186.69 g/mol
Melting Pt: 217 °C
Storage Temperature: Ambient
MDL Number: MFCD00013461
CAS Number: 118427-29-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR