Sarcosine ethyl ester hydrochloride 97%

Synonyms: H-Sar-OEt·HClEthyl 2-(methylamino)acetate hydrochloride

AC30559-0100
200020-648EA 43 USD
200020-648
Sarcosine ethyl ester hydrochloride 97%
Sarcosine ethyl ester hydrochloride
Formula: C₅H₁₁NO₂·HCl
MW: 153.61 g/mol
Melting Pt: 123…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00012506
CAS Number: 52605-49-9
EINECS: 258-037-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR