Thiophene-2-carboxamide 99%

Warning

Synonyms: 2-Thiophenecarboxamide

AC30542-0250 AC30542-0050
103532-216EA 148.5 USD
103532-216 200035-006
Thiophene-2-carboxamide 99%
Thiophene-2-carboxamide
Formula: C₅H₅NOS
MW: 127.17 g/mol
Melting Pt: 179…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00010425
CAS Number: 5813-89-8
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR