Dodecanedioyl dichloride 98%

Danger

AC30534-0250 AC30534-0050
200018-974EA 154 USD
200018-974 200025-218
Dodecanedioyl dichloride 98%
Dodecanedioyl dichloride

Appearance: Clear yellow to brown Liquid

Formula: C₁₂H₂₀Cl₂O₂
Boiling Pt: 140 °C (.5 mmHg)
Density: 1.06 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00012243
CAS Number: 4834-98-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR