2,3-Dimethylhydroquinone 97%

Warning

Synonyms: 1,4-Dihydroxy-2,3-dimethylbenzene

AC30511-0050 AC30511-0250 AC30511-1000
103512-232EA 1312 USD
103512-232 103509-986 200003-142
2,3-Dimethylhydroquinone 97%
2,3-Dimethylhydroquinone
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Melting Pt: 223…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00009997
CAS Number: 608-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR