1-(3-Fluorophenyl)cyclopentanecarboxylic acid ≥98%

Warning

AC30491-0050
200021-464EA 51.8 USD
200021-464
1-(3-Fluorophenyl)cyclopentanecarboxylic acid ≥98%
1-(3-Fluorophenyl)cyclopentanecarboxylic acid
Formula: C₁₂H₁₃FO₂
MW: 208.2288 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800624
CAS Number: 214262-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR