2-(2-Chloro-6-fluorophenyl)ethanol 98%

Warning

Synonyms: 2-Chloro-6-fluorophenethyl alcohol

AC30471-0050
200016-474EA 74.6 USD
200016-474
2-(2-Chloro-6-fluorophenyl)ethanol 98%
2-(2-Chloro-6-fluorophenyl)ethanol

Appearance: Clear colorless Liquid

Formula: C₈H₈ClFO
Storage Temperature: Ambient
CAS Number: 214262-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR