(S)-(+)-3-Hydroxytetrahydrofuran ≥99%

Supplier: ACROS ORGANICS MS

Warning

AC30443-0010 AC30443-0250 AC30443-0050
200021-384EA 33.05 USD
200021-384 103509-972 200008-260
(S)-(+)-3-Hydroxytetrahydrofuran ≥99%
(S)-(+)-3-Hydroxytetrahydrofuran
Formula: C₄H₈O₂
Boiling Pt: 80 °C (15 mmHg)
Density: 1,298
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00064327
CAS Number: 86087-23-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation +15° to +20° (20°C, 589 nm) (c=2, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR