(S)-(+)-4-Phenyl-2-oxazolidinone 99%

Synonyms: (S)-4-Phenyloxazolidin-2-one

AC30385-0050 AC30385-0010
200021-386EA 225 USD
200021-386 200033-478
(S)-(+)-4-Phenyl-2-oxazolidinone 99%
(S)-(+)-4-Phenyl-2-oxazolidinone
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00043396
CAS Number: 99395-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR