Benzhydrylamine 97%

Warning

Synonyms: Aminodiphenylmethaneα-PhenylbenzylamineDiphenylmethanamine

AC30432-0010 AC30432-0250 AC30432-1000
200042-562EA 15.9 USD
200042-562 103509-970 200011-180
Benzhydrylamine 97%
Benzhydrylamine
Formula: C₁₃H₁₃N
MW: 183.25 g/mol
Boiling Pt: 295 °C (760 mmHg)
Melting Pt: 12 °C
Density: 1.06 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008059
CAS Number: 91-00-9
EINECS: 202-032-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR