L-Serinamide hydrochloride 98%

Synonyms: H-Ser-NH₂·HCl(S)-2-Amino-3-hydroxypropanamide hydrochloride

AC30316-2500 AC30316-0010
103512-216EA 20.3 USD
103512-216 200031-736
L-Serinamide hydrochloride 98%
L-Serinamide hydrochloride
Formula: C₃H₉ClN₂O₂
MW: 140.567 g/mol
Melting Pt: 184…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00067560
CAS Number: 65414-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR