Ethyl-3-aminopropanoate hydrochloride 98%

Warning

Synonyms: Ethyl 3-aminopropanoate hydrochloridebeta-Alanine ethyl ester hydrochloride3-Aminopropionic Acid Ethyl Ester HydrochlorideEthyl 3-Aminopropionate HydrochlorideH-β-Ala-OEt·HCl

AC30306-0100
200011-372EA 54 USD
200011-372
Ethyl-3-aminopropanoate hydrochloride 98%
Ethyl-3-aminopropanoate hydrochloride
Formula: C₅H₁₁NO₂·HCl
MW: 153.6 g/mol
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00012909
CAS Number: 4244-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR