D-(+)-Arabitol 99%

Synonyms: D-Arabitol(2R,4R)-Pentane-1,2,3,4,5-pentaol

AC30287-0050 AC30287-0250
103512-192EA 171 USD
103512-192 200011-932
D-(+)-Arabitol 99%
D(+)-Arabitol
Formula: C₅H₁₂O₅
MW: 152.15 g/mol
Melting Pt: 97…105 °C
Storage Temperature: Refrigerator
EINECS: 207-686-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR