3-Methoxyphenethylamine 98+%

Danger

Synonyms: 2-(3-Methoxyphenyl)ethylamine

AC30261-0050 AC30261-0250
200010-002EA 345 USD
200010-002 200010-000
3-Methoxyphenethylamine 98+%
3-Methoxyphenethylamine
Formula: C₉H₁₃NO
Boiling Pt: 118…119 °C (6 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008187
CAS Number: 2039-67-0
EINECS: 218-017-9
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR