Bromotrispyrrolidinophosphonium hexafluorophosphate 97%

Warning

Synonyms: Bromotripyrrolidinylphosphonium hexafluorophosphateBromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)PyBrOP Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate

AC30213-0250 AC30213-0050
200011-486EA 124 USD
200011-486 200018-372
Bromotrispyrrolidinophosphonium hexafluorophosphate 97%
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate
Formula: C₁₂H₂₄BrF₆N₃P₂
MW: 466.2 g/mol
Melting Pt: >100 °C
Storage Temperature: Freezer
MDL Number: MFCD00077412
CAS Number: 132705-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR