DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate 96%

Synonyms: 6-Hydroxy-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid

AC30194-2500
200011-926EA 68.4 USD
200011-926
DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate 96%
DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate
Formula: C₁₀H₁₁NO₃.xH₂O
MW: 193.2 g/mol (anhy)
Melting Pt: 256…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00800582
CAS Number: 76824-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR