(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥97%

Synonyms: D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride

AC30189-2500
200043-094EA 24.7 USD
200043-094
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥97%
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride
Formula: C₁₀H₁₂ClNO₂
Storage Temperature: Ambient
CAS Number: 41220-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR