Ethephon(2-chloroethylphosphonic acid) 90% tech.

Danger

Synonyms: (2-Chloroethyl)phosphonic acid2-Chloroethylphosphonic acidEthephon

AC30163-0010 AC30163-0050
200009-320EA 175 USD
200009-320 200009-322
Ethephon(2-chloroethylphosphonic acid) 90% tech.
Ethephon (2-Chloroethylphosphonic acid)
Formula: C₂H₆ClO₃P
Melting Pt: 74…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00002142
CAS Number: 16672-87-0
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR