1-Aminocyclopropanecarboxylic acid 99%

Warning

Synonyms: 1-Amino-1-cyclopropanecarboxylic acid

AC30142-2500
200004-688EA 118 USD
200004-688
1-Aminocyclopropanecarboxylic acid 99%
1-Aminocyclopropanecarboxylic acid
Formula: C₄H₇NO₂
MW: 101.11 g/mol
Melting Pt: 257 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009944
CAS Number: 22059-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR