Amberjet® 4200-Cl

AC30133-5000 AC30133-0025
200057-068EA 85.05 USD
200057-068 103518-452
Amberjet® 4200-Cl
Amberjet® 4200-Cl
Storage Temperature: Ambient
MDL Number: MFCD00145586
CAS Number: 60177-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR