cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid 98%

Synonyms: cis-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid(1R,2S)-2-(Boc-amino)cyclopentanecarboxylic Acid

AC29958-2500 AC29958-0010
200003-108EA 90.09 USD
200003-108 103511-462
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid 98%
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229.276 g/mol
Melting Pt: 80…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00800539
CAS Number: 136315-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR