Heptakis-6-iodo-6-deoxy-β-cyclodextrin

Synonyms: heptakis-6-iodo-6-deoxy-β-cyclodextrin

AC29901-5000 AC29901-1000
200049-616EA 165.2 USD
200049-616 200037-568
Heptakis-6-iodo-6-deoxy-β-cyclodextrin
Heptakis-6-iodo-6-deoxy-beta-cyclodextrin

Appearance: Off-white Solid

Formula: C₄₂H₆₃I₇O₂₈
MW: 1904.28 g/mol
Melting Pt: 218…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00803290
CAS Number: 30754-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR