(R)-(+)-4-Isopropyl-2-oxazolidinone 98%

Synonyms: (4R)-(+)-4-Isopropyl-2-oxazolidinone(R)-4-Isopropyloxazolidin-2-one

AC29785-0010 AC29785-0250 AC29785-0050
103509-858EA 824 USD
103509-858 200027-312 200004-528
(R)-(+)-4-Isopropyl-2-oxazolidinone 98%
(R)-(+)-4-Isopropyl-2-oxazolidinone
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00075172
CAS Number: 95530-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR