2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid 96%
|
Warning
|
Synonyms:
4-Amino-4-carboxy-2,2,6,6-tetramethylpiperidine-1-oxyl
, 4-Amino-1-oxyl-2,2,6,6-tetramethylpiperidine-4-carboxylic acid
, TOAC
AC30145-2500
AC30145-0010
200003-116EA
192
USD
200003-116
103511-470
2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid 96%
2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid
Formula:
C₁₀H₁₉N₂O₃ MW: 215.273 g/mol Melting Pt: 230 °C Storage Temperature: Freezer |
MDL Number:
MFCD00800575 CAS Number: 15871-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...