β-Ionone (synthetic) 96%

Synonyms: (E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one(E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-onebeta-Ionone

AC29713-0050 AC29713-1000 AC29713-5000
200018-298EA 46.2 USD
200018-298 200042-698 200030-550
β-Ionone (synthetic) 96%
β-Ionone

Appearance: Clear colorless to yellow Liquid

Formula: C₁₃H₂₀O
MW: 192.30 g/mol
Boiling Pt: 126…128 °C (12 mmHg)
Density: 0.944 g/cm³
Flash Pt: 122 °C
Storage Temperature: Ambient
CAS Number: 14901-07-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5215 (20 °C, 589 nm)
GC ≥95 %
Specific gravity (25 °C) 0.941 to 0.947

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR