4-(4-Chlorophenyl)piperidin-4-ol 99+%, crystalline

Warning

Synonyms: 4-(4-Chlorophenyl)-4-hydroxypiperidine

AC29684-0050 AC29684-0010
200041-732EA 27.9 USD
200041-732 200035-562
4-(4-Chlorophenyl)piperidin-4-ol 99+%, crystalline
4-(4-Chlorophenyl)piperidin-4-ol
Formula: C₁₁H₁₄ClNO
MW: 211.69 g/mol
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00006001
CAS Number: 39512-49-7
EINECS: 254-479-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR