Azetidine 98%

Danger

AC29673-0050 AC29673-0010 AC29673-2500
200018-198EA 133 USD
200018-198 103527-398 200003-078
Azetidine 98%
Azetidine
Formula: C₃H₇N
Boiling Pt: 61…62 °C (760 mmHg)
Density: 0.85 g/cm³
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005165
EINECS: 207-963-8
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR