(R)-Glycidyl-3-nitrobenzenesulfonate 98%

Warning

Synonyms: (R)-(-)-Glycidyl nosylate3-Nitrobenzenesulphonic acid (R)-Glycidyl Ester3-Nitrobenzenesulfonic acid (R)-Glycidyl Ester(R)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate(R)-Oxiran-2-ylmethyl 3-nitrobenzenesulfonate

AC29641-0025
200003-070EA 87.7 USD
200003-070
(R)-Glycidyl-3-nitrobenzenesulfonate 98%
(R)-Glycidyl-3-nitrobenzenesulfonate
Formula: C₉H₉NO₆S
MW: 259.23 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013436
CAS Number: 115314-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR