Sodium hydrosulfide hydrate, flakes, pure

Danger

AC29620-0100
102776-404EA 210 USD
102776-404
Sodium hydrosulfide hydrate, flakes, pure
Sodium hydrosulfide hydrate

Appearance: Yellow to orange to green to red to brown Deliquescent flakes

Formula: H₃NaOS
Melting Pt: 55 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
CAS Number: 207683-19-0
UN: 2949
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR