Methanesulfonamide 98%

Warning

Synonyms: MethylsulphonamideMethylsulfonamide

AC29611-0250
200006-932EA 77.9 USD
200006-932
Methanesulfonamide 98%
Methanesulphonamide
Formula: CH₅NO₂S
MW: 95.13 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007940
CAS Number: 3144-09-0
EINECS: 221-553-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR